Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Well Extraction Plate and Plugs
Product Code. 10321455 Shop All Thermo Scientific Products
Click to view available options
Type:
24 Position Extraction Plate Plugs
24 Well Extraction Plate
48 Position Extraction Plate Plugs
48 Well Extraction Plate
Unit Size:
Each
Pack of 24
Pack of 48
Product Code. 10321455
Brand: Thermo Scientific™ 60104235
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these 24 or 48 well extraction plates and plugs with the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold.
Use these 24 or 48 well extraction plates and plugs with Thermo Scientific™ HyperSep™ Universal Vacuum Manifold. This manifold process your samples in solid phase extraction SPE cartridges or 96 well plates.
TRUSTED_SUSTAINABILITY
Specifications
Description 24 Position Extraction Plate Plugs
For Use With (Application) SPE Sample Processing
For Use With (Equipment) 24 Position Well Extraction Plates, Vacuum Manifolds
Product Line HyperSep
Type 24 Position Extraction Plate Plugs