Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Waste Container
Product Code. 13416926 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Positive Pressure Manifold
Unit Size:
Pack of 250
Product Code. 13416926
Brand: Thermo Scientific™ 60104270
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Collect waste from the Thermo Scientific™ HyperSep™ Positive Pressure Manifold using this pre-drilled waste container.
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Waste Container to collect waste from the manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
TRUSTED_SUSTAINABILITY
Specifications
Description Pre-Drilled Waste Container, 1 pk
For Use With (Application) SPE
For Use With (Equipment) Positive Pressure Manifold
Product Line HyperSep
Type Pre-Drilled Waste Container