Learn More
Thermo Scientific™ HyperSep™ HPLC Columns for Online SPE
Product Code. 10733976 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Hypercarb
HyperSep Retain AX
HyperSep Retain CX
HyperSep Retain PEP
Diameter (Metric) Inner:
2.1 mm
3.0 mm
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sorbent Diameter (Metric) Inner Unit Size
10733976 HyperSep Retain AX 3.0 mm Each
10578015 HyperSep Retain PEP 2.1 mm Each
10169044 HyperSep Retain PEP 3.0 mm Each
10199174 HyperSep Retain CX 2.1 mm Each
10629375 HyperSep Retain CX 3.0 mm Each
10547435 HyperSep Retain AX 2.1 mm Each
10240895 HyperSep Hypercarb 2.1 mm Each
10577435 HyperSep Hypercarb 3.0 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 10733976 Supplier Thermo Scientific™ Supplier No. 60312402
Estimated Shipment: 25-11-2026
Clean-up and concentrate target analytes in minutes with these columns combining SPE chemistries with traditional HPLC column fittings and hardware.
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
HyperSep HPLC Columns for Online SPE combine SPE chemistries with traditional HPLC column fittings and hardware.
HyperSep HPLC Columns for Online SPE are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
TRUSTED_SUSTAINABILITY
Specifications
Description HyperSep Retain-AX; 3.0 mm ID
For Use With (Application) Online Solid Phase Extraction
For Use With (Equipment) Conventional HPLC System
Product Line HyperSep
Sorbent HyperSep Retain AX
Type Online HPLC Column
Diameter (Metric) Inner 3.0 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.