Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Mylar™ Pouches (QuEChERS)
Product Code. 15919547 Shop All Thermo Scientific Products
Click to view available options
Description:
4000 mg Anhydrous Magnesium Sulfate, 2000 mg Sodium Chloride, 1500 mg Sodium Citrate Dibasic Dihydrate
50 mL Empty Centrifuge Tubes
6000 mg Anhydrous Magnesium Sulfate, 1500 mg Anhydrous Sodium Acetate
Mylar Pouch: 1200 mg Sodium Chloride
Mylar Pouch: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride
Mylar Pouch: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 1500 mg Sodium Citrate Dibasic Dihydrate
Mylar Pouch: 4000 mg Anhydrous Magnesium Sulfate, 1000 mg Sodium Chloride, 500 mg Sodium Citrate Dibasic Sesquihydrate, 1000 mg Sodium Citrate Tribasic
Mylar Pouch: 4000 mg Sodium Sulfate, 500 mg Anhydrous Magnesium Sulfate
Mylar Pouch: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride
Mylar Pouch: 6000 mg Anhydrous Magnesium Sulfate, 1500 mg Sodium Chloride, 1500 mg Sodium Citrate Dihydrate 750 mg Disodium Citrate Sesquihydrate
Mylar Slim Line Pouch: 500 mg CEC18, SLIM LINE POUCH with 50 mL Centrifuge Tubes
Mylar Slim Line Pouch: 5000 mg Sodium Sulfate
Unit Size:
Pack of 100
Pack of 50
Product Code. 15919547
Brand: Thermo Scientific™ 60105368SP
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE Mylar pouches for QuEChERS extractions.
Save time and achieve more accurate pesticide determinations using Thermo Scientific™ HyperSep™ Dispersive SPE Mylar Pouches. QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Convenient HyperSep Dispersive SPE Mylar Pouches each include 50 pouches of pre-prepared sorbent. Plug seal caps are not included.
HyperSep Dispersive SPE Extraction Products use magnesium sulfate to aid extraction, along with either sodium chloride, sodium citrate, or anhydrous sodium acetate for "base-sensitive" compounds (e.g., folpet or captan).
All HyperSep Dispersive SPE products are pre-prepared and optimized for the method at hand, so you will:
  • Save valuable preparation time
  • Achieve more consistent analytical results due to consistency in products and rigorous quality control
  • Get cleaner extracts and lower peak counts as a result of using highest quality, pre-prepared products
QuEChERS Technique
Developed by the USDA Eastern Regional Research Center in 2003, the QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) technique is increasingly becoming the technique of choice for extraction and clean-up of pesticide residues.
The QuEChERS method is a two-step process: extraction followed by clean-up. There are four variations of the QuEChERS method currently in use:
  • The original QuEChERS method uses sodium chloride to enhance extraction
  • Dispersive AOAC 2007.01 uses sodium acetate as a buffer, replacing sodium chloride
  • The Dual-Phase variation introduces PSA and GCB to remove high levels of chlorophyll and plant sterols in the final extract without the loss of planar pesticides (polar aromatics) using an acetone:toluene solvent mix (3:1)
  • European Version is similar to the AOAC method, except the extraction uses sodium chloride, sodium citrate dihydrate and disodium citrate sesquihydrate instead of sodium acetate
TRUSTED_SUSTAINABILITY
Specifications
Description Mylar Slim Line Pouch: 5000 mg Sodium Sulfate
For Use With (Application) QuEChERS Method
Product Line HyperSep
Sorbent Sodium Sulfate
Type QuEChERS Sample Prep Mylar Pouch