missing translation for 'onlineSavingsMsg'
Learn More

Hyperoside, TRC

Product Code. 30397963
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397963 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397963 Supplier Toronto Research Chemicals Supplier No. TRCH9982005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hyperoside
CAS 482-36-0
Purity Grade Notes HPLC
Molecular Formula C21 H20 O12
Quantity 5 mg
Synonym Hyperin (7CI,8CI), 2-(3,4-Dihydroxyphenyl)-3-(β-D-galactopyranosyloxy)-5,7-dihydroxy-4H-1-benzopyran-4-one, 3,3',4',5,7-Pentahydroxyflavone 3-O-β-D-galactopyranoside, 3-D-Galactosylquercetin, 3-O-β-D-Galactopyranosyl quercetin, 3-O-β-D-galactopyranosylquercetin, Hyperosid, Hyperoside, NSC 407304, Quercetin 3-O-galactopyranoside, Quercetin 3-O-galactoside, Quercetin 3-O-β-D-galactopyranoside, Quercetin 3-O-β-D-galactoside, Quercetin 3-O-β-galactopyranoside, Quercetin 3-O-β-galactoside, Quercetin 3-galactoside, Quercetin 3-β-D-galactoside, Quercetin 3-β-galactoside
InChI Formula InChI=1S/C21H20O12/c22-6-13-15(27)17(29)18(30)21(32-13)33-20-16(28)14-11(26)4-8(23)5-12(14)31-19(20)7-1-2-9(24)10(25)3-7/h1-5,13,15,17-18,21-27,29-30H,6H2/t13-,15+,17+,18-,21+/m1/s1
SMILES OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
Molecular Weight (g/mol) 464.38
Formula Weight 464.0955
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.