Learn More
Plas Labs™ Hypalon Gloves
Product Code. 11817142
Change view
Click to view available options
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11817142 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11817142 Supplier Plas Labs™ Supplier No. 800GH/9/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Plas-Labs™ Hypalon™ Glovebox Gloves are one-piece gauntlets constructed from resistant Hypalon™ chlorosulfonated polyethylene (CSPE) synthetic rubber.
  • White one-piece gauntlet style glovebox gloves
  • Constructed from Hypalon™ chlorosulfonated polyethylene (CSPE) synthetic rubber
  • Resistant to abrasion, oxidising chemicals ozone and UV light
  • Ambidextrous fit
  • Sold in pairs
  • Range of hand sizes and port diameters
Specifications
Color White
Hand Ambidextrous
Material CSM
Product Type Glove Box Gloves
Thickness (English) 0.015 in.
Thickness (Metric) 0.381 mm
For Use With (Equipment) Compatible with most Plas-Lab™ Glove boxes
Port Size (English) 10 in.
Port Size (Metric) 254 mm
Quantity 1 pair
Resistant To Abrasion, oxidising chemicals ozone and UV light