Learn More
Ansell™ Hynit™ 32-815 Series Blue Nitrile Light Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
7
7.5
8
9
10
Unit Size:
12 pairs
Description
- Cut and sewn construction
- Nitrile coated
- Polyester cotton liner
- Blue and white color
Specifications
Specifications
| Back Material | Nitrile |
| Certifications/Compliance | CE Mark, ANSI, EN 388 : 3111 |
| Cuff Material | Nitrile |
| Coating Material | Nitrile |
| Color | Green |
| Exterior Finish | No Texture |
| Hand | Specific |
| Latex Free | Yes |
| Material | Polycotton |
| Product Type | Lightweight Gloves |
| Show More |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur