missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Hygromycin B ReadyMade™ Solution

Product Code. p-200102281
Encompass_Preferred
Click to view available options
Quantity:
20 mL
5 x 20 mL
Unit Size:
1mL
5mL
This item is not returnable. View return policy

Product Code. 30044015

Brand: TOKU-E™ H01120ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hygromycin B ReadyMade™ Solution is provided as a sterile-filtered solution in PBS buffer.

  • CAS Number: 31282-04-9
  • Molecular Formula: C20H37N3O13
  • Molecular Weight: 527.52 g/mol
  • Storage Conditions: 2-8°C
  • Form: Liquid
  • Source: Streptomyces hygroscopicus
  • Spectrum: Hygromycin B is effective against bacteria, fungi, and higher eukaryotic cells
  • Mechanism of Action: Hygromycin B inhibits protein synthesis by strengthening the interaction of tRNA binding in the ribosomal A-site. Hygromycin B also prevents mRNA and tRNA translocation by an unknown mechanism. These are unique mechanisms for an aminoglycoside antibiotic and they differ from the mode of action of neomycin, gentamicin, and G418

Specifications

CAS 31282-04-9
Certifications/Compliance Purity >90% (HPLC)
Chemical Name or Material Hygromycin B
Color Clear, yellowish
Content And Storage 2°C to 8°C
Description Hygromycin B ReadyMade™ Solution 50 mg/mL in PBS buffer
Concentration 50 mg/mL
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in Phosphate-buffered saline (PBS)
For Use With (Application) Microbiology, Plant Biololgy, Eukaryotic Cell Culture Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >90%
Quantity 20 mL
Shipping Condition Ambient
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.