Learn More
Sampling Systems™ Hygienic Butterfly Valve Kit
Product Code. 17912102 Shop All Sampling Systems Products
Change view
Click to view available options
Includes:
Valve with EPDM Seals
Valve with Silicone Seals
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17912102 Valve with Silicone Seals Each
17857817 Valve with EPDM Seals Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17912102 Supplier Sampling Systems™ Supplier No. A13601S
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 23-11-2026
These hygienic butterfly valves are manufactured from 316L stainless steel and are ideal for many applications, including GMP use.
  • Comes with a silicone or EPDM seal
  • Both seals are suitable for food and pharmaceutical use and come with relevant certification
  • Easy to operate: Simply turn the handle to open or close
  • Opening the handle slightly will mean you have reduced product flow
  • Valves are ideal for liquids
Specifications
Product Type Valve Kit
For Use With (Application) Dispensing Liquid
For Use With (Equipment) Drum with Sloping Base
Includes Valve with Silicone Seals
Sterility Non-sterile
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.