missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-949 Microfinished Nitrile-Coated Gloves with Intercept Technology™
GREENER_CHOICE

Product Code. 16099711 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16099711

Brand: Ansell 11949090

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Microfinished nitrile coating enhances grip capabilities while also resisting residual oils and grease. Intercept Technology™ liner provides ANSI Cut Level 2 Protection.

  • Offers cut, abrasion and oil resistance in various dip geometries
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Fortix™ Technology increases abrasion resistance
  • Microfinished nitrile coating enhances grip capabilities
  • Coating is resistant to residual oils and greases present on handled parts
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Hand Specific
Material Intercept Technology Yarn
Size 9
EN388 Standard 4242B
For Use With (Application) Maintenance, Automotive Assembly, Metal Fabrication, and Glass Manufacturing
Glove Cuff Color Black
Glove Cuff Style Knit
Series 11-949
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.