missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-917 Full Nitrile-Coated Seamless Knit Liquid Repellant Gloves
GREENER_CHOICE

Product Code. 16099681 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16099681

Brand: Ansell 11919100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Gloves repel oil, grease, and dirt making them ideal for use in grimey work environments. Seamless nylon liner provides comfort, dexterity, and a secure fit.

  • Liquid repellent against oil, grease, and dirt
  • Outer layer is resistant to polluted objects
  • Seamless nylon liner
  • Suitable for food handling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Exterior Finish Rough
Hand Specific
Latex Free No
Material Nylon
Product Type Mechanical Protection Gloves
Size 10
EN388 Standard 4121A
For Use With (Application) Maintenance, General Handling, Quality Control, and Metal Fabrication
Glove Cuff Color White
Glove Cuff Style Knit
Series 11-919
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.