Learn More
Ansell HyFlex™ 11-730 Polyurethane-Coated Gloves with Intercept™ Technology
Product Code. 16669083 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16669083
Brand: Ansell 11730070
Only null left
Add to basket
Add to basket
Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.
  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 407:2004, CE 0493, EN 420:2003 + A1:2009
Construction Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Cut- and Abrasion-resistant Glove
Size 7
EN388 Standard 4X43D
For Use With (Application) Metalwork, Parts Assembly, and Press Injection Molding
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Lining Lined
Packaging Type Case
Puncture Rating ANSI/ISEA 105-16 Level 3
Series 11-730
Thumb Style Wing
Yarn Gauge 13