Learn More
Ansell HyFlex™ 11-528 Cut-Resistant Polyester Spandex Gloves with Intercept Technology™
Product Code. p-7215056 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16069601
Brand: Ansell 11528060
Only null left
Add to basket
Add to basket
Glass-free intercept liner is skin friendly, and provides ANSI A2 Cut Protection. NBR Foam coating enhances grip capabilities in dry, or light oil applications.
  • Glass-free intercept liner provides increased cut protection
  • Enhanced flexibility for range of motion and tactility when handling small parts
  • Grip performance for safe handling in dry to light oily applications
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • NBR Foam coating provides ANSI 4 Abrasion Resistance
  • DMF-free
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 6
EN388 Standard 3X32B
For Use With (Application) Automotive Assembly, Metal Fabrication, Mining, and Transportation
Gender Unisex
Glove Cuff Style Knit
Series 11-528
Yarn Gauge 18