Description
- Heightened defenses: Designed with INTERCEPT™ Cut Resistance Technology and meeting ANSI/ISEA 105-2024 CUT A5 and EN ISO E standards, HyFlex™ 11-755 industrial gloves protect against lacerations and help prevent common workplace injuries
- Enhanced tactile sensitivity: Palm-dipped, with nylon liner, these PU-coated gloves benefit from heightened tactile sensitivity and touchscreen compatibility
- Elevated comfort: Lightweight and fitted to natural hand contours, HyFlex™ 11-755 PPE gloves cause minimal hand fatigue
- Improved dexterity: These polyurethane gloves' ultralight design also increases flexibility and ease of use
- Certified skin-friendliness: HyFlex™ 11-755 industrial working gloves are Dermatest™-approved for skin-friendliness
Specifications
Specifications
| Coating Material | Polyurethane |
| Color | Gray |
| Cut Resistance Rating | Level E |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Tungsten, Spandex, Polyester, Polyurethane |
| Product Type | Industrial Gloves |
| Size | 7 |