Learn More
Hydroxy Pioglitazone (M-II) (Mixture of Diastereomers), TRC
Product Code. 30398082
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398082 10 mg -
30391265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30398082 Supplier Toronto Research Chemicals Supplier No. TRCH95208010MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101931-00-4
Molecular Formula C19 H20 N2 O4 S
Molecular Weight (g/mol) 372.44
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)-2-hydroxyethoxy]phenyl]methyl]-, 5-[[4-[2-(5-Ethyl-2-pyridyl)-2-hydroxy-ethoxy]phenyl]methyl]thiazolidine-2,4-dione
IUPAC Name 5-[[4-[2-(5-ethylpyridin-2-yl)-2-hydroxyethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CCc1ccc(nc1)C(O)COc2ccc(CC3SC(=O)NC3=O)cc2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C19H20N2O4S/c1-2-12-5-8-15(20-10-12)16(22)11-25-14-6-3-13(4-7-14)9-17-18(23)21-19(24)26-17/h3-8,10,16-17,22H,2,9,11H2,1H3,(H,21,23,24)
Formula Weight 372.1144
Chemical Name or Material Hydroxy Pioglitazone (M-II) (Mixture of Diastereomers)