Learn More
Hydroxy Chlorothalonil, TRC
Product Code. 30008199
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008199 1 g 1g
18012820 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008199 Supplier Toronto Research Chemicals Supplier No. TRCH8250601G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28343-61-5
Molecular Formula C8 H Cl3 N2 O
Molecular Weight (g/mol) 247.47
Synonym Isophthalonitrile, 2,4,5-trichloro-6-hydroxy- (8CI), 2,4,5-Trichloro-6-hydroxy-1,3-benzenedicarbonitrile, 1,3-Dicyano-4-hydroxy-2,5,6-trichlorobenzene, 2,5,6-Trichloro-4-hydroxy-1,3-benzenedicarbonitrile, 4-Hydroxy-2,5,6-trichloroisophthalonitrile, 4-Hydroxychlorothalonil, Hydroxychlorothalonil, SDS 3701, R182281
IUPAC Name 2,4,5-trichloro-6-hydroxybenzene-1,3-dicarbonitrile
SMILES Oc1c(Cl)c(Cl)c(C#N)c(Cl)c1C#N
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8HCl3N2O/c9-5-3(1-12)6(10)7(11)8(14)4(5)2-13/h14H
Formula Weight 245.9154
Percent Purity >95%
Chemical Name or Material Hydroxy Chlorothalonil
Grade HPLC