Learn More
Brannan™ Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate
Product Code. 12899207
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Product Code. 12899207
Brand: Brannan™ 10/602/0
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate, 1.025 to 1.035 x 0.0002 150mm length Brannan
1.025 to 1.035 x 0.0002 150mm length Brannan
Specifications
Type Hydrometer
Length (Metric) 150 mm
For Use With (Application) Specific Gravity Measurement