Description
- The capacity varies depending on the model.
Specifications
Specifications
| Diameter (Metric) | 6.35 cm |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate glass |
| Disposable | Reusable |
| Includes | No Graduation Line |
| Base Type | Flat Circular |
| Capacity (English) | 3.4 fl. oz. |
| Height (English) | 7.3 in. |
| Height (Metric) | 18.5 cm |
| Product Type | Hydrometer Jar |