Learn More
Fisherbrand™ Hydrometer
Click to view available options
Specific Gravity Range:
0 to 100% NaCl Saturation
0 to 26.5% NaCl Saturation
1000 to 2000 g/L
1400 to 1620 g/L
1600 to 1820 g/L
1720 to 1790 g/L
1800 to 2000 g/L
2000 to 2500 g/L
2500 to 3000 g/L
650 to 1000 g/L
795 to 910 g/L
895 to 1000 g/L
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Type | Hydrometer |
| Length (Metric) | 300 mm |
| Brix Range | 0 to 26.5% NaCl Saturation |
| For Use With (Application) | Ideal for harsh environment or field measurement, endure shocks without risk of breakage |
| Specific Gravity Range | 0 to 26.5% NaCl Saturation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.