Learn More
Fisherbrand™ Plastic Baumé Hydrometer, 0-25 Bé°
Click to view available options
Baume Accuracy:
± 0.2 Bé
Unit Size:
Each
Description
- The Baumé scale measures the density of a variety of liquids heavier than water
- Made of shatterproof plastic to eliminate glass breakage
- Made of HACCP compliant plastic that is suitable for use with food
- Scale division of 0.2 Bé, with accuracy of ± 0.2 Bé
- Instrument is single-packed in a transparent square plastic sleeve
Specifications
Specifications
| Quantity | 1Each |
| Type | Hydrometer |
| Length (Metric) | 300 mm |
| Includes | Square plastic sleeve |
| Baume Accuracy | ± 0.2 Bé |
| Baume Divisions | 0.2 Bé |
| Material | Plastic |
| Baume Range | 0 to 25° Bé |
| For Use With (Application) | Ideal for harsh environment or field measurement, endure shocks without risk of breakage |
| Temperature of Standardization | 15°C |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.