Learn More
Esculape Laboratoire™ Hydrogel compresses
Product Code. 11798383
Change view
Click to view available options
Dimensions (L x W):
10 x 10 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11798383 10 x 10 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11798383 Supplier Esculape Laboratoire™ Supplier No. 3170403
  • ONLINE EXCLUSIVE PRICE
    14.67€
    6.93€ / Pack of 1
    Save 7.74€
    52% Off
Estimated Shipment: 09-12-2026
Hydrogel burn compress helps to reduce swelling While providing immediate cooling, relieves pain and moisturises the skin
Hydrogel burn compress helps to reduce swelling While providing immediate cooling, relieves pain and moisturises the skin.
  • Four person, easy to carry first aid kit
  • Soft red pouch with nylon double zipper and belt loop
  • Includes: 1 survival blanket; 2 packs of 5 compresses; 1 roll of tape 5m x 20mm 2 gauze bandages 3m x 70mm; 10 adhesive bandages; 4 fixing clips 3 alcohol compresses; 3 chlorhexidine compresses; 3 anti-blow compresses 3 mosquito compresses; 1 pair of scissors; 1 pair of forceps 2 pairs of vinyl gloves; 1 first aid guide
Specifications
Product Type Hydrogel Compresses
Dimensions (L x W) 10 x 10 cm
For Use With (Application) For Immediate Cooling of Burn