Learn More
Esculape Laboratoire™ Hydrogel Compress
Product Code. 11708393
Change view
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11708393 20 x 20 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11708393 Supplier Esculape Laboratoire™ Supplier No. 3170503
  • ONLINE EXCLUSIVE PRICE
    27.30€
    12.97€ / Pack of 1
    Save 14.33€
    52% Off
Estimated Shipment: 09-12-2026
Sterile hydrogel, suitable for all types of thermal burns
  • Sterile hydrogel compress
  • Size 20 x 20 cm
  • Allows immediate cooling of the burn
  • Relieves pain, interrupts the in-depth development of the burn
  • Reduces edema and promotes local hydration
  • For all types of thermal burns (1st, 2nd and 3rd degree)
Specifications
Sterility Sterile
Dimensions (L x W) 20 x 20 cm
For Use With (Application) All Types of Thermal Burns