Learn More
Macherey-Nagel™ Hydrodex β-6TBDM GC Capillary Column
Click to view available options
Length (Metric):
10 m
25 m
50 m
Unit Size:
Each
Pack of 25
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.1 mm |
| Stationary Phase | Hydrodex β-6TBDM |
| For Use With (Application) | Enantiomer Separation |
| Temperature Range | 230°C to 250°C |
| Length (Metric) | 10 m |
| Packing Material | Hydrodex β-6TBDM |
| Quantity | 1 Each |