missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Hydrocortisone Stock Solution

Product Code. p-7217689
Encompass
Click to view available options
Quantity:
3 mL
10 x 3 mL
Unit Size:
3mL
Pack of 10
This item is not returnable. View return policy

Product Code. 17109921

Brand: STEMCELL Technologies 07926

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrocortisone Stock Solution (200X) for use with PneumaCult™-ALI, MammoCult™, EpiCult™, 10 pack

  • Hydrocortisone Stock Solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines
  • Hydrocortisone Stock Solution is provided at a concentration of 96 μg/mL, which is ideal as a 200X stock solution for use with PneumaCult™-ALI (Catalog #05001), MammoCult™ (Catalog #05620), EpiCult™-B (Human; Catalog #05601), and EpiCult™-C (Catalog #05630) media
  • It may also be used for preparing PneumaCult™-Ex Medium (Catalog #05008) or PneumaCult™-Ex Plus Medium (Catalog #05040)

Specifications

Cell Type Airway Cells, Cancer Cells and Cell Lines, Mammary Cells
For Use With (Application) Cell Culture
Quantity 10 x 3 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.