missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrocinnamic-alpha,alpha-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Phenylpropanoic-2,2-D2 Acid |
| CAS | 19136-97-1 |
| Molecular Formula | C9 D2 H8 O2 |
| Quantity | 0.5 g |
| Synonym | Benzenepropanoic-α,α-d2 acid, Hydrocinnamic-α,α-d2 acid (8 CI), 3-Phenylpropanoic acid-d2, 3-Phenylpropanoic-2,2-d2 acid, 3-Phenylpropanoic-2,2-D2 acid, β-Phenylpropionic-d2 acid, β-Phenylpropionic-α-d2 acid, Hydrocinnamic-d2 acid, Hydrocinnamic acid-d2 |
| InChI Formula | InChI=1 S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5 H,6-7H2,(H,10,11)/i7D2 |
| SMILES | [2 H]C([2 H])(Cc1ccccc1)C(=O)O |
| IUPAC Name | 2,2-dideuterio-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 152.187 |
| Formula Weight | 152.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction