Learn More
Alla France™ Hydrochloric Acid Hydrometer
Description
Order Info
dimLengthMetric
Specifications
Specifications
| Quantity | 1Each |
| Type | Hydrochloric Acid Hydrometer |
| Accuracy | ±0.004g/mL |
| Length (Metric) | 300 mm |
| Specific Gravity Range | 1 to 1.200 g/mL |