Learn More
Hydrochloric Acid 8 M For Laboratory Use and Tritrations, Chem Lab
Product Code. 12675925
Change view
Click to view available options
:
Hydrochloric Acid 8 M For Laboratory Use and Titrations, 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12675925 Hydrochloric Acid 8 M For Laboratory Use and Titrations, 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12675925 Supplier Chem-Lab Supplier No. CL05.0319.1000
Specifications
Color Colorless
CAS 7647-01-0
Melting Point -25°C
Boiling Point 107°C
Concentration 8 M
Density 1.12 g/mL
Packaging Polyethylene
Traceability to NIST Reference standards acc. NIST SRM 84l
Quantity 1 L
Physical Form Liquid
Molecular Weight (g/mol) 36.46
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 8 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid