missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 37% v.p. For Laboratory Use and With DAB, Chem Lab

Product Code. 13248203
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13248203 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13248203 Supplier Chem-Lab Supplier No. CL00.0319.1000
  • ONLINE EXCLUSIVE PRICE
    28.00€
    18.90€ / 1 L
    Save 9.10€
    32% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7647-01-0
Melting Point -25°C
Boiling Point 50°C
Density 1.19 g/mL
Non_volatile Matter < 0.005%
Packaging Glass bottles
Residue after Ignition < 0.002%
Assay Percent Range > 37%
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
Grade Very Pure
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 37% v.p.
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.