Learn More
Emma Safety Footwear Hydro-Tec™ Stability Pro Inlay Soles
Product Code. 11340590
Change view
Click to view available options
Size:
44
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11340590 44 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11340590 Supplier Emma Safety Footwear Supplier No. MM00109044
  • ONLINE EXCLUSIVE PRICE
    42.49€
    27.78€ / Pack of 1
    Save 14.71€
    34% Off
Estimated Shipment: 28-12-2026
Prevents fallen arches and gives extra support to the inside of the foot.
  • Provides extra support to inside of foot
  • Prevents fallen arches
Specifications
Color Gray
For Use With (Application) Provides extra support to inside of foot
For Use With (Equipment) Prevents fallen arches
Size 44
Product Type Stability Pro Inlay Soles