Learn More
Lauda™ Hydro H 20 SOW Shaking Water Bath with CEE7/7 Plug
Product Code. 16659345
Change view
Click to view available options
Temperature Range:
25°C to 80°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Unit Size
16659345 25°C to 80°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16659345 Supplier Lauda™ Supplier No. L002910
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Shaking water bath for exactly reproducible temperatures
  • Easy-to-read LED display
  • Setting and displaying temperature values in increments of 0.1°C
  • Membrane keypad with concise symbols
  • Digital setpoint adjustment
  • Speed continuously adjustable, load-independent, with gently controlled start-up
  • 1-point temperature controller
  • over temperature protection for operation with non-flammable liquids and water deficiency protection
  • Electronic monitoring of temperature controller
  • Cooling coil as standard
  • Shaking device with a gentle start-up and overload-protected
  • Shaking device is guided on corrosion-resistant special ball bearings
  • Orbital shaking motion
  • Shaking basket can be easily removed for cleaning purposes
  • Cover frame with condensate channel
  • Bath vessel with drain valve
  • Heat insulating lid made of stainless steel, double-walled with inner curvature prevents condensate from dripping back into vessels
  • Bath interior, cover, shaking basket and heating element made of stainless steel
  • Lid and perforated tray as standard
  • Technical Features:
    • Working temperature range: 25 to 80 °C
    • Ambient temperature range: 10 to 40 °C
    • Temperature stability: 0.1 ± K
    • Heater power max: 1.5 kW
    • Power consumption max: 1.5 kW
    • Power consumption: 10 A
    • Size of bath (W x D x H): 450 x 300 x 160 mm
    • Overall dimensions (W x D x H): 635 x 505 x 400 mm
    • Shaking amplitude: 14 mm
    • Power supply: 230 V, 50/60 Hz
    • Power plug: power cord with angled plug (CEE7/7)
  • Specifications
    Temperature Range 25°C to 80°C
    Voltage 230 V