missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-1,1-d2 Alcohol-OD, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Ethanol-O,1,1-D3 |
| CAS | 22544-42-9 |
| Molecular Formula | C2 D3 H3 O |
| Quantity | 0.5 g |
| Synonym | Ethan-1,1-d2-ol-d (9 CI), Ethyl-1,1-d2 alcohol-d (6 CI, 8 CI), Ethanol-O,1,1-d3, O,1,1-Trideuterioethanol, Ethanol-O,1,1-D3, Ethanol-d3, Ethyl-1,1-d2 Alcohol-OD |
| InChI Formula | InChI=1 S/C2H6O/c1-2-3/h3H,2H2,1H3/i2D2,3 D |
| SMILES | [2 H]OC([2 H])([2 H])C |
| IUPAC Name | 1,1-dideuterio-1-deuteriooxyethane |
| Molecular Weight (g/mol) | 49.0869 |
| Formula Weight | 49.0607 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction