Learn More
Juchheim Laborgeraete™ Hydraulic Laboratory Jack
Product Code. 10444501
Change view
Click to view available options
:
200 x 230mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10444501 200 x 230mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10444501 Supplier Juchheim Laborgeraete™ Supplier No. 02297/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these hydraulic lifting stages for easy, safe lifting. Juchheim Laborgeraete™ Hydraulic Laboratory Jacks feature robust, light aluminum alloy construction.
  • Complete with pedal and anti-sag device
  • Aluminum alloy construction
  • Stable, parallel movement
Specifications
Description Hydraulic Laboratory Jack
Dimensions (L x W x H) 230 x 200 x 91 mm
For Use With (Application) Easy, safe lifting
Height (Metric) 91 mm (min.), 285 mm (max.)
Length (Metric) 230 mm
Load Bearing Capacity (Metric) 25 kg
Width (Metric) 200 mm