Learn More
Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media
Click to view available options
Quantity:
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188, Sodium Bicarbonate
None
Without Additives:
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
No Phenol Red, No HEPES
Product Type:
PF CHO LS
PF-CHO LS
PF-CHO MPS
Unit Size:
10 L
1000mL
5 L
Pack of 6
Description
Specifications
Specifications
| Form | Liquid |
| Product Type | PF CHO LS |
| With Additives | Glutamine, Poloxamer 188, Sodium Bicarbonate |
| Without Additives | No Phenol Red, No HEPES |
| Quantity | 6 x 1000 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2° to 8°C |