Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum
Product Code. 17682075
Change view
Click to view available options
Quantity:
25 to 40 Doses
30 to 45 Doses
Unit Size:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17682075 25 to 40 Doses 1 L
17692075 30 to 45 Doses 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17682075 Supplier Cytiva Supplier No. SH30401.02IH2540
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
  • Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
    • Sourced from animals that are typically less than 10 days old
    • Traceable
    • Conforms to USDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -10°C or Lower, Once Thawed Store at 2°C to 8°C
Country of Origin New Zealand
Product Type Newborn Calf Bovine Serum
Quantity 25 to 40 Doses
Volume (Metric) 1000 mL
For Use With (Application) Enhance Growth of Cells