Learn More
Cytiva HyClone™ Minimal Essential Medium with Earle's (MEM/EBSS), Powder
Product Code. 11551821
Click to view available options
Quantity:
10 L
2 x 5 L
50 L
With Additives:
Glutamine
Glutamine, Non Essential Amino Acids (NEAA)
Without Additives:
No α-Thioglycerol
No Sodium Bicarbonate
None
Unit Size:
10 L
50 L
Pack of 2
Product Code. 11551821
Brand: Cytiva SH30050.03
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Minimal Essential Medium with Earle's (MEM/EBSS), manufactured using ISO 9001:2000-certified processes.
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Product Type MEM/EBSS with L-glutamine, nonessential amino acids (NEAA)
Sterilization Method Non-sterile
With Additives Glutamine, Non Essential Amino Acids (NEAA)
Without Additives No Sodium Bicarbonate
Quantity 10 L
Certifications/Compliance cGMP and ISO 9001:2000
Content And Storage 2° to 8°C, away from light and moisture
Description MEM/EBSS with L-Glutamine, Nonessential Amino Acids