Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
Product Code. 17236946
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
None
Unit Size:
1 L
1000mL
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
17236946 1000 mL Irradiated (25 to 40 doses) 1 L
10761274 500 mL None 500mL
12330043 500 mL Irradiated 500mL
11582830 1000 mL None 1000mL
12694217 1000 mL Heat Inactivated 1000mL
12684217 1000 mL Irradiated 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17236946 Supplier Cytiva Supplier No. SH30626.03IR2540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A cost-effective alternative to Fetal Bovine Serum (FBS).
  • Ensured safety against bovine diseases
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Ensured uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
    Buffer Lactate Dehydrogenase
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin New Zealand
    Form Liquid
    Additive Alkaline Phosphatase
    Product Type Calf Bovine Serum
    Quantity 1000 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing Irradiated (25 to 40 doses)
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Heat Inactivated No
    Product Line HyClone
    Mycoplasma Detected Yes
    USDA Safety Testing Included Yes