Learn More
Cytiva HyClone™ Insect Cell Culture Media
Product Code. p-7058064
Click to view available options
Quantity:
1000 mL
500 mL
Unit Size:
1000mL
500mL
Product Code. 10288023
Brand: Cytiva SH30065.01
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Increase cell yield with multiple insect lines in a variety of culture environments with HyClone™ Insect Cell Culture Media.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 500 mL
Content And Storage 2° to 8°C
Description PETE