Learn More
Cytiva HyClone™ IMDM Media, Powder
Product Code. 11918075
Change view
Click to view available options
Quantity:
2 x 5 L
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11918075 2 x 5 L Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11918075 Supplier Cytiva Supplier No. SH30005.02
  • ONLINE EXCLUSIVE PRICE
    51.18€
    63.09€ / Pack of 2
    Save POA
    -23.27% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Specifications
Form Powder
Product Type IMDM with L-glutamine, HEPES; without alpha-thioglycerol
Sterilization Method Non-sterile
With Additives Glutamine, HEPES
Without Additives No α-Thioglycerol
Quantity 2 x 5 L
Certifications/Compliance cGMP and ISO 9001:2000
Content And Storage 2° to 8°C, away from light and moisture
Description IMDM with L-Glutamine, HEPES, without Alpha-Thioglycerol