Learn More
Cytiva HyClone™ Hank's 1X Balanced Salt Solutions
Product Code. 10617394
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
Unit Size:
1000mL
100mL
500mL
Pack of 6
With Additives:
Calcium, Magnesium
Calcium, Magnesium, Phenol Red
Phenol Red
Phenol Red, Calcium, Magnesium
Product Code. 10617394
Brand: Cytiva SH30588.01
Edge
Buffer cell culture growth with 1X balanced salt solutions
  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.4
Quantity 500 mL