Learn More
Cytiva HyClone™ Cell Boost™ 4 Supplement
Artikelnummer. 11586331
Ansicht ändern
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11586331 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11586331 Supplier Cytiva Supplier No. SH30857.01
  • ONLINE EXCLUSIVE PRICE
    281.00€
    255.73€ / 100g
    Save 25.27€
    8% Off
 Restricted Item
Estimated Shipment: 30-11-2026