Learn More
Cytiva HyClone™ CDM4CHO™ Media, Liquid
Product Code. 10433142
Change view
Click to view available options
Quantity:
1000 mL
1000 mL bottle
6 x 1000 mL
With Additives:
Glutamine, Sodium Bicarbonate, Poloxamer 188
Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Glutamine, No Phenol Red, No HEPES, No Protein
No Phenol Red, No HEPES, No Protein
Unit Size:
1000mL
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Unit Size
10433142 1000 mL bottle Glutamine, Sodium Bicarbonate, Poloxamer 188 No Phenol Red, No HEPES, No Protein 1000mL
10298023 6 x 1000 mL Sodium Bicarbonate, Poloxamer 188 No Glutamine, No Phenol Red, No HEPES, No Protein Pack of 6
10463522 1000 mL Sodium Bicarbonate, Poloxamer 188 No Glutamine, No Phenol Red, No HEPES, No Protein 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10433142 Supplier Cytiva Supplier No. SH30557.02
  • ONLINE EXCLUSIVE PRICE
    108.00€
    75.46€ / 1000mL
    Save 32.54€
    30% Off
View all promotions
 Restricted Item
In Stock
Support growth of CHO cell culture. HyClone™ CDM4CHO™ Media, Liquid is a serum-free, chemically defined media.
  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type CDM4CHO Liquid Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Phenol Red, No HEPES, No Protein
Quantity 1000 mL bottle
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description CDM4CHO media