Learn More
Cytiva HyClone™ Bovine Calf Serum, US Origin
Product Code. 16665735
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Product Code. 16665735
Brand: Cytiva SH30072.02NI
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A cost-effective alternative to Fetal Bovine Serum (FBS)
  • Collected by venipuncture from formula-fed veal calves
  • Age of animal at time of collection approximately 16-24 weeks
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Content And Storage ≤-10°C
Age 16 to 22 weeks
Country of Origin United States
Quantity 100 mL
Sterilization Method Sequential-filtered, 100 nm
Source Formula-Fed Veal Calves
Tested For Virus Panel Testing According to 9 CFR 113.53