Learn More
Cytiva HyClone™ ActiSM Powdered Cell Culture Media
Product Code. 17225906
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17225906 1 L 1 L
17215906 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17225906 Supplier Cytiva Supplier No. SH31038.06
  • ONLINE EXCLUSIVE PRICE
    91.90€
    76.06€ / 1 L
    Save 15.84€
    17% Off
 Restricted Item
Estimated Shipment: 24-12-2026
A chemically defined ADCF cell culture medium optimized for high-yield protein production in CHO cell culture processes.
HyClone ActiSM medium is intended for use in combination with HyClone ActiPro medium and Cell Boost 7a and Cell Boost 7b supplements. This medium is part of the ActiPro system of media and supplements designed with adaptation and production in mind. HyClone ActiSM and the ActiPro system are suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
  • Chemically defined ADCF formulation optimized for high-yield protein production
  • Does not contain hypoxanthine, thymidine, and glycine (HAT supplement)
  • Recommended for first step when adapting cells cultured in other lean cell culture media
TRUSTED_SUSTAINABILITY
Specifications
Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine
Quantity 1 L
Content And Storage 2 to 8°C, away from light and moisture