Learn More
MP Biomedicals™ Hyaluronidase, From Streptomyces hyalurolyticus
Shop All MP Biomedical Products
Click to view available options
Quantity:
4 x 100 U
Inhibitors:
Mn2+, Hg2+, K+ Ferricyanide
Molecular Weight (g/mol):
91000
Unit Size:
400 units
Description
- From Streptomyces hyalurolyticus
- Lyophilized, salt-free
- Activity: ∽2000 units/mg
- Protease and lysozyme free
- Each vial contains 100U
Specifications
Specifications
| Concentration | ≈2000 U/mg |
| Color | White |
| pH | 6 |
| For Use With (Application) | Cell biology and analysis |
| Content And Storage | 0°C to 5°C |
| Source | Streptomyces hyalurolyticus |
| Quantity | 4 x 100 U |
| Product Type | Hyaluronidase |
| Molecular Weight (g/mol) | 91000 |
| Inhibitors | Mn2+, Hg2+, K+ Ferricyanide |
| Show More |