missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Hyaluronan DuoSet ELISA

Product Code. 30021635
Change view
Click to view available options
Format:
96-well Microplate
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
30021635 96-well Microplate 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021635 Supplier R&D Systems™ Supplier No. DY361405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Kit components listed are available separately:

  • PBS: (Catalog # DY006), or 137 mM NaCl, 2.7 mM KCl, 8.1 mM Na2HPO4, 1.5 mM KH2PO4, pH 7.2 - 7.4, 0.2 μm filtered
  • Wash Buffer: (Catalog # WA126), or 0.05% Tween™ 20 in PBS, pH 7.2-7.4
  • Reagent Diluent: (Catalog # DY004), or 5% Tween 20 in PBS, 7.2-7.4, 0.2 μm filtered
  • Substrate Solution: 1:1 mixture of Color Reagent A (H2O2) and Color Reagent B (Tetramethylbenzidine) (Catalog # DY999)
  • Stop Solution: 2 N H2SO4 (Catalog # DY994) Microplates: R&D Systems (Catalog # DY990)
  • Plate Sealers: ELISA Plate Sealers (Catalog # DY992)

Hyaluronan DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

This DuoSet ELISA Development kit contains the basic components required for the development of sandwich ELISAs to measure natural Hyaluronan. The suggested diluent is suitable for the analysis of most cell culture supernate samples. Diluents for complex matrices, such as serum and plasma, should be evaluated prior to use in this DuoSet.

Hyaluronan, also known as hyaluronic acid (HA) or sodium hyaluronate, is a naturally occurring linear polymer. Hyaluronan is a glycosaminoglycan (GAG) that is ubiquitously present in the extracellular matrix of all vertebrates and is also present inthe capsule of some strains of Streptococci. Mammalian hyaluronan is synthesized by one of three distinct hyaluronan synthases (HAS1, 2, and 3), which produce HA polymers with different chain lengths and differ in their rates of synthesis. Whereas high molecular weight HA (> 500 kDa) is anti-angiogenic, anti-inflammatory and immunosuppressive, low molecular weight HA (10 - 500 kDa) is highly angiogenic and pro-inflammatory. HA oligomers are anti-apoptotic and upregulate heat shock protein expression. Functionally, hyaluronan molecules are important for the maintenance of a highly hydrated extracellular matrix in tissues, which is involved in cell adhesion and supports cell migration.

The Hyaluronan DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies multi-species Hyaluronan.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Solid Phase Sandwich ELISA
Species Human
Binding Target Interferon gamma
Assay Range 0.37 to 90 pg/mL
Format 96-well Microplate
Sample Type Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma
Sample Volume 100 μL
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.