Learn More
Ansell Edmont™ Hy-Care™ 62-200 Series Blue Natural Rubber Latex Gloves
Shop All Ansell Products
Click to view available options
:
Size: 9
Specifications
Specifications
| Back Material | Latex |
| Cuff Material | Natural Latex |
| Coating Material | Natural Latex |
| Latex Free | No |
| Product Type | Gloves |
| Style | 62-200 |
| Glove Cuff Color | Blue |
| Liner Material | Cotton Jersey |
| Quantity | 12 Pairs |
| Resistant To | Excellent heat or cold insulation |
| Show More |