Learn More
Hamilton™ HVCX Large Body Ceramic Valves
Product Code. 11573494
Change view
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
4-5 Model
8-5 Model
For Use With:
PSD/2, 3 Syringe Drive Module
PSD/4, 8 Syringe Drive Module
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Model For Use With unitSize
11573494 Aluminum 4-5 Model PSD/2, 3 Syringe Drive Module Each
11593444 Stainless Steel 4-5 Model PSD/2, 3 Syringe Drive Module Each
11557884 Stainless Steel 8-5 Model PSD/2, 3 Syringe Drive Module Each
11583434 Aluminum 4-5 Model PSD/4, 8 Syringe Drive Module Each
11583494 Aluminum 8-5 Model PSD/4, 8 Syringe Drive Module Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11573494 Supplier Hamilton™ Supplier No. 650002
Only null left
Add to basket
Add to basket
Designed to provide superior performance with salt solutions.
  • Rigid ceramic fluid path is resistant to scratching from salt crystals
  • Provides superior life and performance
  • Exceptionally easy to service and modify
  • Maximizes flexibility and value
  • Ideal for low pressure applications
Specifications
Description 4-5 Model, 4 Ports, Aluminum Housing
For Use With PSD/2, 3 Syringe Drive Module
Material Aluminum
Model 4-5 Model
Type Ceramic Large Body Valve