Learn More
Hamilton™ HVC Standard Ceramic Valves
Click to view available options
Material:
Aluminum
Stainless Steel
Model:
3-2 Model
3-5 Model
T 3-2 Model
Unit Size:
Each
Description
- Rigid ceramic fluid path is resistant to scratching from salt crystals
- Provides superior life and performance
- Exceptionally easy to service and modify
- Maximizes flexibility and value
- Ideal for low pressure applications
Specifications
Specifications
| Description | 3-5 Model, 3 Ports, Aluminum Housing |
| For Use With | PSD/4 and 8 Syringe Drive Modules |
| Material | Aluminum |
| Model | 3-5 Model |
| Type | Ceramic HV Valve |