missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZSCAN1 Partial ORF (NP_872378, 315 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number NP_872378
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 284312
Molecular Weight (g/mol) 35.97kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16112737
View Documents Promotion Details
Abnova™
H00284312-Q01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16102737
View Documents Promotion Details
Abnova™
H00284312-Q01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

Sequence: REEGPFPCPECGKVFLHNSVLTEHGKIHLLEPPRKKAPRSKGPRESVPPRDGAQGPVAPRSPKRPFQCSVCGKAFPWMVHLIDHQKLHTAHGH
Specifications

Specifications

NP_872378
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ33779/MGC104472/MZF-1
ZSCAN1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
284312
ZSCAN1 (Human) Recombinant Protein (Q01)
REEGPFPCPECGKVFLHNSVLTEHGKIHLLEPPRKKAPRSKGPRESVPPRDGAQGPVAPRSPKRPFQCSVCGKAFPWMVHLIDHQKLHTAHGH
RUO
ZSCAN1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications