missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF81 Partial ORF (NP_009068.1, 1 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00347344-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122867

  • 545.00€ / 25µg

Estimated Shipment: 04-10-2022
to see stock.
Add to basket

Description

Description

Sequence: MPANEDAPQPGEHGSACEVSVSFEDVTVDFSREEWQQLDSTQRRLYQDVMLENYSHLLSVGFEVPKPEVIFKLEQGEGPWTLEGEAPHQSCSDGKFGIKPSQRRISGKSTFHSEMEGEDTLC
Specifications

Specifications

NP_009068.1
Liquid
347344
ZNF81 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ44367/HFZ20/MRX45
ZNF81
Yes
wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
39.16kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MPANEDAPQPGEHGSACEVSVSFEDVTVDFSREEWQQLDSTQRRLYQDVMLENYSHLLSVGFEVPKPEVIFKLEQGEGPWTLEGEAPHQSCSDGKFGIKPSQRRISGKSTFHSEMEGEDTLC
RUO
ZNF81
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers